Learn More
Medchemexpress LLC Isoguvacine (hydrochloride) | 68547-97-7 | 99.8% | 163.60 g/mol | C6H10ClNO2 | 100 MG

Supplier: Medchemexpress LLC HY100810100MG
Isoguvacine hydrochloride is a selective GABAA receptor agonist used in neuroscience research to modulate inhibitory GABAergic signaling. It is supplied as a white to light yellow solid or as a 10 mM solution in DMSO. Intended for laboratory research use only; handle and store according to the product SDS.
- Selective GABAA receptor agonist.
- High purity (99.8%).
- Molecular weight 163.60 g/mol.
- Available as solid and 10 mM DMSO solution.
- White to light yellow appearance.
- Store sealed, away from moisture; follow SDS for solvent storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.