Learn More
Medchemexpress LLC Isohomovanillic acid | 1131-94-8 | 182.18 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0021101G
Encompass_Preferred
Isohomovanillic acid is a product of catecholamine metabolism. Also known as 3-Hydroxy-4-methoxyphenylacetic acid, it can be isolated from urine and is utilized in Parkinson's disease research.
  • Product of catecholamine metabolism
  • Can be isolated from urine
  • Used in Parkinson's disease research
Catalog No. 50-004-47482