Learn More
Medchemexpress LLC Isohomovanillic acid | 1131-94-8 | 182.18 | 5 G

Supplier: Medchemexpress LLC HYW0021105G
Isohomovanillic acid (3-Hydroxy-4-methoxyphenylacetic acid) is a product of catecholamine metabolism. It can be isolated from urine and used in the research of Parkinson's disease.
- Product of catecholamine metabolism
- Can be isolated from urine
- Used in Parkinson's disease research
- Store powder at -20°C for 3 years, or 4°C for 2 years
- Store in solvent at -80°C for 6 months, or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.