Learn More
Medchemexpress LLC Isojacareubin | 50597-93-8 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN83061MG
Encompass_Preferred
Isojacareubin can be isolated from *Hypericum japonicum*. It is classified as a flavonoid (flavone) and appears as a light yellow to yellow solid. This compound is for research use only and not sold to patients.
  • Covalently inhibits SARS-CoV-2 3CLpro
  • Exhibits anti-helicobacter activity
  • Inhibits PKC
  • Suppresses hepatocellular carcinoma metastasis
  • Induces apoptosis
Catalog No. 50-004-19727