Learn More
Medchemexpress LLC Isojacareubin | 50597-93-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN83065MG
Encompass_Preferred
Isojacareubin, a compound isolated from Hypericum japonicum, demonstrates significant biological activities including covalent inhibition of SARS-CoV-2 3CLpro and anti-helicobacter properties. It also functions as a PKC inhibitor, capable of suppressing hepatocellular carcinoma metastasis and inducing apoptosis.
  • Isolated from Hypericum japonicum
  • Covalently inhibits SARS-CoV-2 3CLpro
  • Possesses anti-helicobacter activity
  • Inhibits PKC
  • Suppresses hepatocellular carcinoma metastasis
  • Induces apoptosis
Catalog No. 50-004-17421