Learn More
Medchemexpress LLC (+)-Isolariciresinol | 548-29-8 | MFCD07784475 | 97.3% | 360.40 g·mol-1 | C20H24O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145795MG
Encompass_Preferred
Isolariciresinol is a phenolic lignan with reported anti-inflammatory activity, used in biochemical and pharmacological research including studies of rheumatic inflammation. The compound is supplied in small research pack sizes and is typically provided at high purity. Follow recommended storage conditions to preserve stability and activity.
  • Phenolic lignan with reported anti-inflammatory activity.
  • Suitable for biochemical and pharmacological research applications, including rheumatic inflammation studies.
  • High purity (≈97.3%).
  • Available in small research pack sizes (5 MG and others).
  • Store at 4°C and protect from light; in solvent, store at -80°C up to 6 months.
Catalog No. 50-002-21422