Learn More
Cayman Chemical IsoleucIn-d10 10mg
SDP
Supplier:  Cayman Chemical 3484210 MG
Encompass_Preferred
An internal standard for the quantification of isoleucine by GC- or LC-MS
Catalog No. 50-291-2658