Learn More
Selleck Chemical LLC isoleucine S3752-1g
SDP
Supplier:  Selleck Chemical LLC S37521G
Encompass_Preferred
Isoleucine (Ile) is a non-polar uncharged (at physiological pH) aliphatic amino acid that is used in the biosynthesis of proteins
Catalog No. 50-310-9982