Learn More
Selleck Chemical LLC Isoliensinine S9247-1MG
SDP
Supplier:  Selleck Chemical LLC S92471MG
Encompass_Preferred
Isoliensinine is an alkaloid produced by the edible plant Nelumbo nucifera It possesses anti-cancer anti-fibrosis anti-proliferative antioxidant and anti-inflammatory activities
Catalog No. 50-000-25262