Learn More
Medchemexpress LLC Isolongifolene | 1135-66-6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN736310MM/1ML
Encompass_Preferred
Isolongifolene is a tricyclic sesquiterpene isolated from Murraya koenigii. It attenuates Rotenone-induced oxidative stress, mitochondrial dysfunction, and apoptosis by regulating PI3K/AKT/GSK-3β signaling pathways.
  • Antioxidant properties
  • Anti-inflammatory properties
  • Anticancer properties
  • Neuroprotective properties
  • Attenuates Rotenone-induced cytotoxicity and apoptosis in SH-SY5Y neuroblastoma cells
Catalog No. 50-004-17432