Learn More
Medchemexpress LLC Isolongifolene | 1135-66-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN736310MG
Encompass_Preferred
Isolongifolene is a tricyclic sesquiterpene isolated from Murraya koenigii. It attenuates Rotenone-induced oxidative stress, mitochondrial dysfunction, and apoptosis by regulating PI3K/AKT/GSK-3β signaling pathways. This compound possesses antioxidant, anti-inflammatory, anticancer, and neuroprotective properties.
  • Attenuates Rotenone-induced oxidative stress, mitochondrial dysfunction, and apoptosis
  • Regulates PI3K/AKT/GSK-3β signaling pathways
  • Possesses antioxidant properties
  • Possesses anti-inflammatory properties
  • Possesses anticancer properties
  • Possesses neuroprotective properties
Catalog No. 50-004-19232