Learn More
Medchemexpress LLC Isolongifolene | 1135-66-6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN7363
Encompass_Preferred
Isolongifolene is a tricyclic sesquiterpene isolated from Murraya koenigii. It has demonstrated antioxidant, anti-inflammatory, anticancer, and neuroprotective properties. It is intended for research use only.
  • Attenuates rotenone-induced oxidative stress, mitochondrial dysfunction, and apoptosis.
  • Regulates PI3K/AKT/GSK-3β signaling pathways.
  • Shows efficacy in SH-SY5Y neuroblastoma cells.
  • Down-regulates Bax, caspases-3, 6, 8, and 9 expression.
  • Up-regulates Bcl-2 expression.
  • Regulates p-P13K, p-AKT, and p-GSK-3β expression.
Catalog No. 50-004-21877