Learn More
TARGETMOL CHEMICALS INC Isolongifolene 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T116855MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Isolongifolene ((-)-Isolongifolene), a tricyclic sesquiterpene isolated from Murraya koenigii, exhibits antioxidant, anti-inflammatory, anticancer, and neuroprotective properties. It mitigates Rotenone-induced oxidative stress, mitochondrial dysfunction, and apoptosis by modulating the PI3K/AKT/GSK-3beta signaling pathways. Purity 99.31%
Catalog No. 50-288-0571