Learn More
TARGETMOL CHEMICALS INC Isomaltose 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0581200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Isomaltose (D-Isomaltose) is a reducing disaccharide, similar to maltose, but with an alpha-(1-6)-linkage instead of the alpha-(1-4)-linkage. Purity 98.27%
Catalog No. 50-288-0716