Learn More
Cayman Chemical IsomItraphylIn 10mg
SDP
Supplier:  Cayman Chemical 3945210 MG
Encompass_Preferred
An alkaloid with anticancer activity; antagonist of Drosophila ecdysteroid receptors in a reporter assay using Drosophila BII cells (EC50 = 160 µM); reduces the proliferation of, and induces apoptosis in, CCRF CEM C7H2 leukemia cells at 50 or 100 µM
Catalog No. 50-291-7022