Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isomogroside V | 1126032-65-2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN68155MG

Encompass_Preferred

Isomogroside V is a sweetener extracted from *Siraitia grosvenorii* (Swingle), also known as Monk Fruit. This product is supplied as a solid, white to off-white in appearance, with a high purity of 99.91%. It is classified as a triterpene and is used in various applications requiring a natural sweetener.

  • Extracted from *Siraitia grosvenorii* (Swingle)
  • Purity of 99.91%
  • Solid appearance, white to off-white
  • Sweetener properties
  • Molecular formula C60H102O29
  • Molecular weight 1287.43

Catalog No. 50-004-17949


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.