Learn More
Medchemexpress LLC Isonicotinamide-d4 | 1219799-40-2 | 99.9% | 126.15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW012843S50MG
Encompass_Preferred
Isonicotinamide-d4 is the deuterium labeled Isonicotinamide. It is intended for research use only and not for sale to patients. Stable heavy isotopes like deuterium have been incorporated into drug molecules for use as tracers in drug development, and deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54875