Learn More
Medchemexpress LLC Isonicotinamide-d4 | 1219799-40-2 | 99.91% | 126.15 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW012843S10MG
Encompass_Preferred
Isonicotinamide-d4 is the deuterium labeled Isonicotinamide. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-55047