Learn More
Medchemexpress LLC HY-I0736 100mg Medchemexpress, Isonicotinic acid CAS:55-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYI0736100MG
Encompass_Preferred
Medchemexpress, HY-I0736 100mg Isonicotinic acid CAS:55-22-1 Isonicotinic acid is a metabolite of Isoniazid. Isoniazid is converted to Isonicotinic acid by hydrazinolysis, with the Isoniazid to Isonicotinic acid biotransformation also to be catalyzed by cytochrome P450 (CYP) enzymes, e.g., CYP2C. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0204
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.