Learn More
Cayman Chemical IsopentedronhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1156310 MG
Encompass_Preferred
A variant of pentedrone in which the α-propyl and β-keto groups have switched positions; intended for forensic and research purposes
Catalog No. 50-295-2506