Learn More
Medchemexpress LLC Isopimaric acid | 5835-26-7 | 5 MG

Supplier: Medchemexpress LLC HYN34635MG
Isopimaric acid is a potent opener of large conductance calcium-activated K+ (BK) channels, derived from plants, particularly Pinaceae Pinus nigra Arn. This product is a white to off-white solid with a molecular weight of 302.45 and a chemical formula of C20H30O2. It is classified as an "Other Terpenoid" with a purity of 99.88% and is intended for research use only.
- Potent opener of large conductance calcium activated K+ (BK) channels
- Intended for research use only
- Appears as a white to off-white solid
- Molecular weight: 302.45
- Chemical formula: C20H30O2
- Classified as a terpenoid
- Initial source: Plants (Pinaceae Pinus nigra Arn)
- Purity: 99.88%
- Solubility: ≥ 25 mg/mL (82.66 mM) in DMSO
- Storage: Powder at -20°C for 3 years; In solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.