Learn More
Indofine Chemical Isopimpinellin, 98%, 25 Mg
SDP
Supplier:  Indofine Chemical 19661 25 MG
Encompass_Preferred
ISOPIMPINELLIN 25 MG, Coumarin, 4,9-Dimethoxypsoralen, C13H10O5, MW: 246.22, 98+%, 482-27-9, MFCD00017407
Catalog No. 50-908-649