Learn More
Apexbio Technology LLC Isoprenaline HCl(Synonyms: Isoproterenol hydrochloride, Isoprenaline hydrochloride, Isadrine), 1g, CAS: 51-30-9.
SDP
Supplier:  Apexbio Technology LLC B13361000
Encompass_Preferred
Isoprenaline hydrochloride (CAS 51-30-9) is a synthetic sympathomimetic amine functioning as a non-selective -adrenoceptor agonist structurally analogous to epinephrine Its mechanism involves the stimulation of both 1- and 2-adrenergic receptors resulting in increased cardiac output through enhanced chronotropic and inotropic effects alongside bronchial smooth muscle relaxation Due to these pharmacological properties isoprenaline hydrochloride is utilized in research exploring cardiac arrhythmias conduction disorders bradycardia and bronchospasm
Catalog No. 50-000-60161