Learn More
Medchemexpress LLC Isoprenaline hydrochloride | 51-30-9 | 99.86% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB04685G
Encompass_Preferred
Isoprenaline (Isoproterenol) hydrochloride is a non-selective, orally active β-adrenergic receptor agonist. It possesses potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities. It can be utilized for research pertaining to bradycardia and bronchial asthma.
  • Non-selective β-adrenergic receptor agonist
  • Potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities
  • Utilized for research related to bradycardia and bronchial asthma
  • Increases particulate cGMP- and cilostamide-inhibited, low-Km cAMP phosphodiesterase activity
  • Inhibits insulin-stimulated glucose transport activity in rat adipocytes
  • Increases cyclic AMP levels
  • Increases the transcriptional activity of the Gi alpha-2 gene
Catalog No. 50-004-07725