Learn More
Cayman Chemical IsoprInosIn 10mg
SDP
Supplier:  Cayman Chemical 1649210 MG
Encompass_Preferred
A complex of acetaminobenzoic acid, dimethylaminoisopropanol, and inosine in a 3:3:1 ratio; has a number of immunomodulatory effects, including inducing T-lymphocyte differentiation, augmenting macrophage and NK cell functions, and stimulating IL-2 production; shows antiviral activity
Catalog No. 50-297-0484