Learn More
Medchemexpress LLC Isopromethazine hydrochloride | 5568-90-1 | MFCD01706895 | 50mg
SDP
Supplier:  Medchemexpress LLC HYZ909950MG
Encompass_Preferred
Isopromethazine hydrochloride is an impurity in the pharmaceutical preparations of Promethazine (HY-B1296) Promethazine is an N-substituted phenothiazine with the actions and the uses of the antihistamines (H1-receptor antagonists)[1]
Catalog No. 50-002-69422