Learn More
Medchemexpress LLC Isopropamide (iodide) | 71-81-8 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB166750MG
Encompass_Preferred
Isopropamide iodide is a long-acting quaternary anticholinergic agent. It is used in peptic ulcer and other gastrointestinal disorders characterized by hyperacidity and hypermotility.
  • Acts as a long-acting quaternary anticholinergic agent
  • Targets mAChR
  • Functions within GPCR/G Protein and neuronal signaling pathways
  • Aids in managing peptic ulcer conditions
  • Supports treatment of gastrointestinal disorders
  • Helps alleviate hyperacidity
  • Contributes to reducing hypermotility
Catalog No. 50-004-12133