Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Propanol | 67-63-0 | MFCD00011674 | 1l
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 593041LF
Encompass
2-Propanol | Purity: 99.5% | Mol Wt: 60.1 | 67-63-0 | MFCD00011674 | 1l
Catalog No. NC1085483