Learn More
Medchemexpress LLC Isopropyl (1-((5-chloro-8-hydroxyquinolin-7-yl)methyl)piperidin-4-yl)carbamate | 1928763-08-9 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12269710MM 1ML
Encompass_Preferred
ML418 is a potent, selective potassium channel blocker used for research on Kir7.1. Supplied as high-purity powder or as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for in vitro screening and CNS pharmacology studies.
  • Potent Kir7.1 potassium channel blocker.
  • High purity (≈99.8%) supporting reliable biological data.
  • Available as powder and as a 10 mM DMSO solution for convenience.
  • Molecular weight 377.87 and formula C19H24ClN3O3.
  • Store powder and solutions under recommended cold conditions to preserve stability.
Catalog No. 50-002-30697