Learn More
Medchemexpress LLC Isopropyl benzenesulfonate | 6214-18-2 | MFCD04038771 | 100.0% | 200.25 g/mol | C9H12O3S | 25 G

Supplier: Medchemexpress LLC 25GHYW043437
Isopropyl benzenesulfonate is a sulfonate ester supplied as a reagent and analytical reference material, noted for its role as a potential genotoxic impurity.
- cas number 6214-18-2.
- Chemical formula C9H12O3S.
- Molecular weight 200.25 g/mol.
- Purity 99.99%.
- Physical appearance colorless to light yellow liquid.
- Pack size 25 g, with larger quantities available by quote.
- Used as an analytical standard and impurity reference in chemical analysis.
- Handle with appropriate safety precautions due to potential genotoxicity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.