Learn More
Medchemexpress LLC Isoprothiolane | 50512-35-1 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB185810MM/1ML
Encompass_Preferred
Isoprothiolane is a blast fungicide that exhibits antifungal, anti-inflammatory, and insecticidal properties, primarily acting on fungi during the penetration and growth stages of infecting hyphae. It can also reduce serum phospholipid and total lipid concentrations, regulating lipid metabolism, and is utilized in fatty liver research.
  • Suppresses *Nilaparvata lugens* and *Sogatella furcifera* populations
  • Promotes cell proliferation and inhibits IL-1 and IL-6 production in bovine mammary epithelial cells
  • Inhibits 14C-acetate incorporation into fatty acids and triglycerides
  • Inhibits 14C-methionine incorporation into phospholipids
  • Used as an insecticide, pesticide, and in research related to fatty liver
Catalog No. 50-004-11787