Learn More
Medchemexpress LLC Isoquinoline, 3,4-dihydro-1-phenyl- | 52250-50-7 | MFCD00006760 | ≥96.0% | 207.28 | 10 G

Supplier: Medchemexpress LLC HY3213610G
1-Phenyl-3,4-dihydroisoquinoline is a chemical reagent. It has a molecular weight of 207.28 and typically appears as a light yellow to brown liquid. This product is suitable for various laboratory applications.
- Purity of at least 96.0%
- Molecular weight of 207.28 g/mol
- Appears as a light yellow to brown liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.