Learn More
Medchemexpress LLC HY-N2082 5mg Medchemexpress, Isorhamnetin 3-O-galactoside CAS:6743-92-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20825MG
Encompass_Preferred
Medchemexpress, HY-N2082 5mg Isorhamnetin 3-O-galactoside CAS:6743-92-6 Isorhamnetin 3-O-galactoside (Cacticin), a flavonoid glycoside isolated from Artemisia capillaris Thunberg, which ameliorates CCl4-induced hepatic damage by enhancing the anti-oxidative defense system and reducing the inflammatory signaling pathways. Isorhamnetin 3-O-galactoside (Cacticin) has antithrombotic and anti-inflammatory activities[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0364