Learn More
Medchemexpress LLC HY-N0777 5mg Medchemexpress, Isorhamnetin-3-O-glucoside CAS:5041-82-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07775MG
Encompass_Preferred
Medchemexpress, HY-N0777 5mg Isorhamnetin-3-O-glucoside CAS:5041-82-7 Isorhamnetin-3-O-glucoside, a natural compound widely contained in many vegetables and rice, could be metabolized in intestinal microbiota after digestion [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3712