Learn More
Cayman Chemical IsorhamntIn 10mg
SDP
Supplier:  Cayman Chemical 1649610 MG
Encompass_Preferred
A natural flavonol aglycone that has antioxidant activity and inhibits xanthine oxidase (IC50 = 0.40 µM); competitively inhibits MATE1, Ki = 0.32 µM); potentiates the neurological actions of nerve growth factor, diminishes the cardiotoxic impact of doxorubicin, and can have beneficial anti-cancer effects
Catalog No. 50-296-9870