Learn More
Cayman Chemical IsorhamntIn 3OnohesperIdo 10mg
SDP
Supplier:  Cayman Chemical 3460410 MG
Encompass_Preferred
A flavonoid glycoside with antioxidant and osteoclastogenic activities; inhibits XO and scavenges superoxide radicals in a cell-free assay (IC50s = 48.75 and 30 UG/ml, respectively); promotes RANKL-induced osteoclastogenesis of mouse bone marrow-derived macrophages
Catalog No. 50-294-9433