Learn More
TARGETMOL CHEMICALS INC ISORHAPONTIGENIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T381410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isorhapontigenin (Isorhapotogenin) is a tetrahydroxylated stilbenoid with a methoxy group. It is an isomer of rhapontigenin and an analog of resveratrol. It is found in the Chinese herb Gnetum cleistostachyum in Gnetum parvifolium and in the seeds of the palm Aiphanes aculeata. purity: 99%
Catalog No. 50-258-0113