Learn More
Selleck Chemical LLC Isorhychophylline S9310-1MG
SDP
Supplier:  Selleck Chemical LLC S93101MG
Encompass_Preferred
Isorhynchophylline (IsoRhy 7-Isorhyncophylline) is a major tetracyclic oxindole alkaloid isolated from the Chinese herbal medicine Uncaria rhynchophylla (Miq )Jacks (Gouteng in Chinese) It acts as a neuronal autophagy inducer with therapeutic potential for cardiovascular and central nervous system diseases
Catalog No. 50-000-24472