Learn More
Medchemexpress LLC Isorosmanol | 93780-80-4 | 1 MG

Supplier: Medchemexpress LLC HYN41911MG
Isorosmanol is an abietane-type diterpene isolated from Rosmarinus officinalis L. aerial parts, with antioxidant, neuroprotective, and neurotrophic effects. Isorosmanol inhibits AChE activity and melanin synthesis.
- High purity: 99.40%
- Molecular weight: 346.42
- Molecular formula: C20H26O5
- Appearance: Solid
- Color: White to off-white
- Target: Cholinesterase (ChE)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.