Learn More
Selleck Chemical LLC Isosakuranetin S9438-1MG
SDP
Supplier:  Selleck Chemical LLC S94381MG
Encompass_Preferred
Isosakuranetin (4 -Methylnaringenin) an O-methylated flavonoid is the 4 -methoxy derivative of naringenin a flavanone found in Citrus species
Catalog No. 50-000-24881