Learn More
Medchemexpress LLC Isosakuranin | 491-69-0 | MFCD29049499 | 100.0% | 448.42 g·mol⁻¹ | C22H24O10 | 10 MG

Supplier: Medchemexpress LLC HYN429610MG
Isosakuranin is a naturally occurring flavonoid (flavanone glycoside) isolated from the fruits of Paliurus ramosissimus. It is supplied as a high-purity research reagent for in vitro studies and should be stored protected from light.
- High purity: 99.97%.
- Molecular formula C22H24O10 and molecular weight 448.42 g·mol⁻¹.
- Suitable for in vitro research applications and biochemical studies.
- Store protected from light at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
- Available in mg-scale quantities for laboratory research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.