Learn More
Medchemexpress LLC Isosakuranin | 491-69-0 | MFCD29049499 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN429620MG
Encompass_Preferred
Isosakuranin is a natural product derived from the fruits of *Paliurus ramosissimus*. It exhibits anti-inflammatory activity against LPS-stimulated mouse RAW264.7 cells by decreasing PGE2 production.
  • Derived from *Paliurus ramosissimus*
  • Exhibits anti-inflammatory activity against LPS-stimulated mouse RAW264.7 cells
  • Decreases PGE2 production
  • IC50 value in RAW264.7 cells is > 50 μM
  • Purity of 99.97%
  • Appears as a white to off-white solid
  • Classified as a flavonoid and flavanone
  • Originates from plants such as *Aconitum jaluense Kom.*, *Citrus trifoliata L.*, and *Rhamnaceae*
Catalog No. 50-004-19599