Learn More
Medchemexpress LLC Isoscoparin-2′′O-glucoside | 97605-25-9 | 1 MG

Supplier: Medchemexpress LLC HYN51411MG
Isoscoparin-2′′O-glucoside is a flavonoid found in the yellow grain mutant of rice (Oryza sativa L.). It exhibits antioxidant activity. It has a molecular weight of 624.54 and a chemical formula of C28H32O16. It appears as a light yellow to yellow solid.
- Found in the yellow grain mutant of rice
- Exhibits antioxidant activity
- Molecular weight: 624.54
- Chemical formula: C28H32O16
- Purity: 99.27%
- Appearance: Light yellow to yellow solid
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.