Learn More
Medchemexpress LLC HY-N0779 5mg Medchemexpress, Isosilybin CAS:72581-71-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07795MG
Encompass_Preferred
Medchemexpress, HY-N0779 5mg Isosilybin CAS:72581-71-6 Isosilybin (Isosilybinin) is a flavonoid from milk thistle; inhibits CYP3A4 induction with an IC 50 of 74 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3713