Learn More
Apexbio Technology LLC Isosorbide 652-67-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B17745.1
Encompass_Preferred
Isosorbide exerts its biological activity primarily through osmotic effects increasing renal water and electrolyte excretion and thereby promoting diuresis Based on these pharmacological properties isosorbide holds research potential in fluid management studies renal function modulation and in the investigation of physiological responses to altered water-electrolyte balance Additionally it serves as a molecular precursor or intermediate in pharmaceutical synthesis and biomaterial research particularly for the development of biodegradable polymers and sustained drug-delivery systems
Catalog No. 50-000-94561