Learn More
Medchemexpress LLC Isosorbide (D-Isosorbide; Dianhydro-D-glucitol) | 652-67-5 | 98.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB146910MM/1ML
Encompass_Preferred
Isosorbide (D-Isosorbide) is an orally active vasodilating agent that can be used for the research of heart failure and angina (chest pain). It is also an oral hyperosmotic diuretic.
  • Orally active vasodilating agent.
  • Useful for the research of heart failure and angina (chest pain).
  • Functions as an oral hyperosmotic diuretic.
Catalog No. 50-004-11217