Learn More
Cayman Chemical Isosorbide dinitrate 50MG
SDP
Supplier:  Cayman Chemical 2399050 MG
Encompass_Preferred
An NO donor; induces vasodilation in isolated precontracted rabbit femoral vein and artery (IC50s = 0.56 and 1.82 µM, respectively); decreases oxygenation of tumor tissue in vivo in mice; inhibits platelet aggregation in vitro and in vivo
Catalog No. 50-293-3291