Learn More
Medchemexpress LLC Isosorbide dinitrate | 87-33-2 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB140910MM/1ML
Encompass_Preferred
Isosorbide dinitrate (ISDN) is an NO donor that prevents LV remodeling and degradation of cardiac function following myocardial infarction (MI). It is for research use only.
  • Prevents LV remodeling
  • Prevents degradation of cardiac function
  • Improves pulmonary artery pressure
  • Exhibits cardioprotective effect against myocardial ischemia-reperfusion injury
  • Solid appearance
  • White to off-white color
  • High purity
Catalog No. 50-004-12910