Learn More
Medchemexpress LLC Isosorbide mononitrate | 16051-77-7 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB064210MM/1ML
Encompass_Preferred
Isosorbide mononitrate is an orally active nitric acid compound used for angina pectoris by dilating blood vessels and lowering blood pressure. It increases the viability and proliferation of HUVECs by decreasing apoptosis and elevating the expressions of vegf, kdrl, pdgfr in zebrafish embryos. It is promising for research of heart failure and coronary heart disease.
  • Dilates blood vessels and lowers blood pressure
  • Increases HUVECs viability and proliferation
  • Decreases HUVECs apoptosis
  • Elevates expressions of vegf, kdrl, and pdgfr in zebrafish embryos
  • Inhibits angiogenesis, tumor growth, and metastasis
  • Suitable for research of heart failure and coronary heart disease
Catalog No. 50-004-09176