Learn More
Cayman Chemical IsothIpendylhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 335825 MG
Encompass_Preferred
An antihistamine; protects against MES-induced seizures in rats (ED50 = 57.9 mg/kg)
Catalog No. 50-294-8101